Brandschutzzeichen
Feuerlöschgerät, PVC, selbstklebend, 200×200 mm
Artikelnr. 116-2016
Anbieter: KROSCHKE
Springe zu:
Product Details & Documents
These self-adhesive, PVC film signs are used to indicate fire equipment. According to EC Directive 92/58/EEC and related national legislation on safety and/or health signs and also according to to VBG 125 (a regulation of the German employer's liability insurance association), signboards/signs for fire equipment must be permanently fitted and clearly recognisable at all times.
- Suitable for use inside and outside
- Visible from a distance of ∼12 metres
- Long-lasting persistent luminescence material available
Zertifizierung: VBG 125 (now known as BGV A8).
Achtung! Bitte beachten: Long-lasting persistent luminescence fire signs must be displayed in new and refurbished buildings if no safety lighting is available.
Spezifikationen
- Bezeichnung:Feuerlöschgerät
- Abmessungen:200×200 mm
- Material:PVC, selbstklebend