AuftragsverfolgungKontakt
Electrode Plug Cable Combinations | Avantor
Electrode Plug Cable Combinations
Plug cable combinations, electrode socket, 5 m two core cable up to 80 °C, free end, 5 m
Artikelnr. SCOI285124654
Anbieter:   SI Analytics
Electrode Plug Cable Combinations
Artikelnr. SCOI285124654
Anbieter:  SI Analytics
Electrode Plug Cable Combinations
Electrode Plug Cable Combinations
Plug cable combinations, electrode socket, 5 m two core cable up to 80 °C, free end, 5 m
Preis
68,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Electrode cables along with electrode socket, 1 m two core cable up to 80 °C, for resistance thermometer and 2-pole-conductivity cell without temperature sensor. Available with different cable length.
Spezifikationen
  • Bezeichnung:
    Plug cable combinations, electrode socket, 5 m two core cable up to 80 °C, free end
  • Länge:
    5 m
  • Seq:
    03