Ringlichter
Ringlicht, ununterbrochen, mit flexibler Länge ca. 1.000 mm, Innen-Ø 66 mm, min. Arbeitsabstand 25 mm
Artikelnr. 631-0033
Anbieter: SCHOTT AG Lighting and imaging
Springe zu:
Product Details & Documents
Ring lights are used to provide intense, uniform and shadow-free illumination.
Dokumente
Spezifikationen
- Bezeichnung:Ringlicht, ununterbrochen, mit flexibler Länge ca. 1.000 mm, Innen-Ø 66 mm, min. Arbeitsabstand 25 mm
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.