Express Eingabe
Austria
AuftragsverfolgungKontakt
Cyclohexan, BAKER, Laborreagens, J.T.Baker®

Cyclohexan, BAKER, Laborreagens, J.T.Baker®

Glass bottle, 1 L
Catalog # 8104.1000
Flammable
Health
Irritant
Environment
Supplier:   Avantor
Cyclohexan, BAKER, Laborreagens, J.T.Baker®
Catalog # 8104.1000
Supplier:  Avantor
CAS Number:  110-82-7
Cyclohexan, BAKER, Laborreagens, J.T.Baker®
Cyclohexan, BAKER, Laborreagens, J.T.Baker®

Glass bottle, 1 L

CAS Number:  110-82-7
Price
69,40 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0,7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Raumtemperatur
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Assay (by GC):
    min. 99 %
  • Residue after Evaporation:
    max. 0,005 %
  • Water (H2O):
    max. 0,02 %

Specifications

  • Packung:
    Glass bottle
  • VE:
    1 L

Specifications