n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Glasflasche, 1 L
Anbieter: Avantor
n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Artikelnr. 8044.1000
Anbieter: Avantor
CAS Nummer: 110-54-3
Springe zu:
Product Details & Documents
Synonyme: Hexyl hydride, Hexanen, Hexane
- CAS Nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL-Nummer:MFCD00009520
- Molekulargewicht:86.18 g/mol
- Merck-Index:13,04712
- Siedepunkt:69 °C (1013 hPa)
- Schmelzpunkt:–94,3 °C
- Dichte:0,659 g/cm³ (20 °C)
- Blitzpunkt:–22 °C
- Lagertemperatur:Raumtemperatur
- EINECS:203-777-6
- UN:1208
- ADR:3,II
Spezifikationen
Spezifikation Testergebnisse
- Assay (by GC):min. 95 %
- Aromatic Compounds (as C6H6):max. 0,02 %
- Color (APHA):max. 10
- Density (g/ml) at 25°C:max. 0,657
- Residue after Evaporation:max. 0,001 %
- Sulfur Compounds (as S):max. 0,005 %
- Thiophene:passes test
- Water Soluble Titrable Acid (meq/g):max. 0,0003
- Trace Impurities (in ppm):-
- Aluminium (Al):max. 0,5
- Barium (Ba):max. 0,1
- Boron (B):max. 0,02
- Cadmium (Cd):max. 0,05
- Calcium (Ca):max. 0,5
- Chromium (Cr):max. 0,02
- Cobalt (Co):max. 0,02
- Copper (Cu):max. 0,02
- Iron (Fe):max. 0,1
- Lead (Pb):max. 0,1
- Magnesium (Mg):max. 0,1
- Manganese (Mn):max. 0,02
- Nickel (Ni):max. 0,02
- Tin (Sn):max. 0,1
- Zinc (Zn):max. 0,1
Spezifikationen
- Packung:Glasflasche
- VE:1 L
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



