Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Staining baskets and racks

Staining basket, for 30 horizontal slides, 172×86 mm
  631-9740
 :   DELTALAB
Staining baskets and racks
  631-9740
 :  DELTALAB
Staining baskets and racks

Staining basket, for 30 horizontal slides, 172×86 mm

 
149,00 €
   
1 * 1 ST
 

Staining baskets and racks, made of stainless steel. Designed for 76×26 mm slides.

  • Good chemical resistance
  • Good temperature resistance
 
  • Bezeichnung:
    Staining basket, for 30 horizontal slides, 172×86 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.