Extraction thimbles
40×123 mm, 2,0 mm
Anbieter: MACHEREY-NAGEL
Springe zu:
Product Details & Documents
Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.
- Fat determination in food and feed
- Determination of binder content in asphalt
- Analysis of soil contaminants
- Determination of additives in rubber and elastomers
Spezifikationen
- I-Ø×H:40×123 mm
- Seq:30
- Stärke:2,0 mm