Calciumhypochlorit, tech.
Glass bottle for solids, 250 g
Anbieter: Thermo Fisher Scientific
Calciumhypochlorit, tech.
Artikelnr. 42548.30
Anbieter: Thermo Fisher Scientific
CAS Nummer: 7778-54-3
Springe zu:
Product Details & Documents
CAS No.: 7778-54-3 Molecular Formula: CaCl2O2 Formula Weight: 142.98 Physical Form: Granular Density: 2.350 Appearance: White to pale brown Odour: Like chlorine pH: 11,5 Melting Point: 100° Merck Reference: 14,1674 UN No.: 1748 MDL No.: MFCD00010900
DeOdourant, oxidizing agent, bleaching agent
- CAS Nummer:7778-54-3
- Formel:CaCl₂O₂
- MDL-Nummer:MFCD00010900
- Molekulargewicht:142.98 g/mol
- Merck-Index:14,1674
- Schmelzpunkt:100 °C
- Dichte:2,350
- Lagertemperatur:Raumtemperatur
- EINECS:231-908-7
- UN:1748
- ADR:5.1,II
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:250 g



