Graphite electrodes, crater
Graphite electrode, crater-drillpoint/undercut, 4,57×38,10 mm
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Graphite electrodes are used mainly in electric arc furnace steel production.
- AGKSP grade
- Ultra F purity
- Insoluble in water
They are also used to refine steel in ladle furnaces and in other smelting processes, furnance design, sharp requirements, water spray rings and charging practice.
Dokumente
Spezifikationen
- Ø×L:4,57×38,10 mm
- Bezeichnung:Graphite electrode, crater-drillpoint/undercut
- Seq:01