Kolophonium
Glass bottle for solids, 250 g
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
CAS No.: 8050-09-7 Physical Form: Powder/solid Solubility: Soluble in alcohol, benzene, ether. Insoluble in water Merck Reference: 14,8266 MDL No.: MFCD00132205
Synonyme: Rosin, gum, Rosin, Gum rosin
- CAS Nummer:8050-09-7
- MDL-Nummer:MFCD00132205
- Merck-Index:14,8266
- Schmelzpunkt:75…81 °C
- Dichte:1,1 g/cm³ (20 °C)
- Blitzpunkt:188 °C
- Lagertemperatur:Raumtemperatur
- EINECS:232-475-7
Dokumente
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:250 g



