n-Decan ≥99%
Glasflasche für Feststoffe, 100 g
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Purity: 99+% CAS No.: 124-18-5 Molecular Formula: C10H22 Formula Weight: 142.29 Flash Point: 46°(115°F) Physical Form: Liquid Density: 0.730 Refractive Index: 1.4115 Appearance: Colourless Odour: Petrol-like Melting Point: -30° Boiling Point: 172-174° Beilstein Reference: 1696981 UN No.: 2247 MDL No.: MFCD00008954
- CAS Nummer:124-18-5
- Formel:H₃C(CH₂)₈CH₃
- MDL-Nummer:MFCD00008954
- Molekulargewicht:142.28 g/mol
- Siedepunkt:172…174 °C
- Schmelzpunkt:–30 °C
- Dichte:0,730
- Blitzpunkt:46 °C (115 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:204-686-4
- UN:2247
- ADR:3,III
Documents
Spezifikationen
Spezifikationen
- Packung:Glasflasche für Feststoffe
- VE:100 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



