Ethylpropionat
Glasflasche für Feststoffe, 1 kg
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
CAS No.: 105-37-3 Molecular Formula: CH3CH2CO2CH2CH3 Formula Weight: 102.13 Flash Point: 12°(54°F) Physical Form: Liquid Density: 0.891 Refractive Index: 1.3850 Melting Point: -73° Boiling Point: 99° Beilstein Reference: 506287 Merck Reference: 14,3847 UN No.: 1195 MDL No.: MFCD00009308
Synonyme: Propionic acid ethyl ester
- CAS Nummer:105-37-3
- Formel:C₅H₁₀O₂
- MDL-Nummer:MFCD00009308
- Molekulargewicht:102.13 g/mol
- Merck-Index:14,3847
- Siedepunkt:99 °C
- Schmelzpunkt:–73 °C
- Dichte:0,891
- Blitzpunkt:12 °C (54 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:203-291-4
- UN:1195
- ADR:3,II
Documents
Spezifikationen
Spezifikationen
- Packung:Glasflasche für Feststoffe
- VE:1 kg
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



