Graphite electrodes, counter
Graphite electrode, counter-flat top, 3,05×38,10 mm
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Graphite electrodes are used as homogeneous catalysts.
- AGKSP grade
- Ultra F purity
- Light sensitive and hygroscopic
- Incompatible with oxidising agents
- Applications include piston rings, thrust bearings, journal bearings and vanes
These are widely used in electric arc furnaces (EAF) and ladle furnaces (LF) for the melting of steel, non-ferrous metals and some non-metallic minerals. Carbon based seals are used in the shafts and fuel pumps of many aircraft jet engines.
Dokumente
Zertifikate
Suche nach Chargennummermsds_ALFA014746 msds_ALFA014751 msds_ALFA014754
Sicherheitsdatenblatt (SDS)
Spezifikationen
- Ø×L:3,05×38,10 mm
- Bezeichnung:Graphite electrode, counter-flat top
- Seq:01