Tellurdioxid, tech.
Kunststoffflasche für Feststoffe, 250 g
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
CAS No.: 7446-07-3 Molecular Formula: TeO2 Formula Weight: 159.60 Physical Form: Powder Density: 5.8 Odour: Odourless Merck Reference: 14,9123 MDL No.: MFCD00011263
- CAS Nummer:7446-07-3
- Formel:O₂Te
- MDL-Nummer:MFCD00011263
- Molekulargewicht:159.6 g/mol
- Merck-Index:14,9123
- Siedepunkt:1245 °C (1013 hPa)
- Schmelzpunkt:733 °C
- Dichte:5,8
- Lagertemperatur:Raumtemperatur
- EINECS:231-193-1
Spezifikationen
Spezifikationen
- Packung:Kunststoffflasche für Feststoffe
- VE:250 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



