Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Retort rods, with M10 thread

16 mm, 1000 mm, M10
Artikelnr. BOCH5116
Anbieter:   BOCHEM
Retort rods, with M10 thread
Artikelnr. BOCH5116
Anbieter:  BOCHEM
Retort rods, with M10 thread
Retort rods, with M10 thread

16 mm, 1000 mm, M10

Preis
61,90 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Stainless steel 18/10.

  • Rods for stand bases
  • M10 thread
Spezifikationen
  • Ø:
    16 mm
  • Gewinde:
    M10
  • Länge:
    1000 mm
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.