Serological pipettes
10 ml, 0,1 ml, +, Orange, 25 Stück/Packung, 500 Stück/Karton
Anbieter: SARSTEDT
Springe zu:
Product Details & Documents
PS, graduated, sterile.
- Plugged
- Easy identification by colour code
- Larger pipetting volume due to negative graduation
- Optimised mouth piece for universal fit in most common pipettors
- Easy to open, static-free packaging
Dokumente
Spezifikationen
- Farbcode:Orange
- Verpackt:25 Stück/Packung, 500 Stück/Karton
- Steril:+
- Kapazität:10 ml
- Teilung:0,1 ml