AuftragsverfolgungKontakt
Stative
With detachable legs, 120×210 mm
Catalog # 451-9106
Supplier:   RSG
Stative
Catalog # 451-9106
Supplier:  RSG
Stative
Stative
With detachable legs, 120×210 mm
Price
43,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Separable tripods made from stainless steel.
Specifications
  • I-Ø×H:
    120×210 mm
  • Bezeichnung:
    With detachable legs
  • Seq:
    02
Accessories
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • RSGA497.370
    Tripod extensions, set of three, total height up to 370 mm
    Log in for availability
    1 * 3 ST - 78,80 €