Pincettes
Blunt, 250 mm
Artikelnr. 232-0092
Anbieter: USBECK Laborgeräte
Springe zu:
Product Details & Documents
Stainless steel. A large range of quality forceps for various laboratory applications, production applications etc.
- Practical
- Anti-magnetic
- Resistant to corrosives
Dokumente
TDS USBE3016.pdf USBE3010_TDS_EN USBE3012_TDS_EN USBE3013_TDS_EN USBE3015_TDS_EN
Technisches Datenblatt
Spezifikationen
- Typ:Blunt
- Länge:250 mm