AuftragsverfolgungKontakt
Big blue | Avantor
Big blue
Transpack packaging extract, 100 reactions, 1
Artikelnr. STRA200221
Marketsource
Anbieter:   STRATAGENE
Big blue
Artikelnr. STRA200221
Anbieter:  STRATAGENE
Marketsource
CAS Nummer:  
Big blue
Transpack packaging extract, 100 reactions, 1
CAS Nummer:  
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
These are used in all Big Blue rodents and cell lines.
Transpack packaging extract is optimised for use with Big Blue Rodents and Cell lines as well as the lambda CII select Assay.
The Big Blue transgenic rodent mutagenesis assay is an in vivo system for assessing the genetic toxicity of test compounds.
The Lambda Select-cII mutation detection system for Big Blue rodents is a positive mutant-screening method for mutagenesis studies using Big Blue transgenic rodents.
Big Blue paque colour controls with four separate phage suspensions of colour mutants mixed with non-mutant phage.
Spezifikationen
Spezifikationen
  • VE:
    1
  • Bezeichnung:
    Transpack packaging extract
  • Tests pro Kit:
    100 reactions
Spezifikationen