n-Hexan, EMPLURA®, Supelco®
Edelstahltrommel, Mehrweg, 190 L
Anbieter: Merck
Springe zu:
Product Details & Documents
EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.
Synonyme: Hexyl hydride, Hexanen, Hexane
- CAS Nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL-Nummer:MFCD00009520
- Molekulargewicht:86.18 g/mol
- Merck-Index:13,04712
- Siedepunkt:69 °C (1013 hPa)
- Schmelzpunkt:–94,3 °C
- Dichte:0,659 g/cm³ (20 °C)
- Blitzpunkt:–22 °C
- Lagertemperatur:Raumtemperatur
- UN:1208
- ADR:3,II
- :M-Clarity™ Qualitätsstufe = MQ200
- EINECS:203-777-6
Documents
Zertifikate
Suche nach Chargennummerexternalurl.pdf ProductPage_104368
Product specification
msds_104368
SDS
Spezifikationen
Spezifikationen
- Packung:Edelstahltrommel, Mehrweg
- VE:190 L
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



