Express Eingabe
Austria
AuftragsverfolgungKontakt
Cyclohexan, EMPLURA®, Supelco® | Avantor

Cyclohexan, EMPLURA®, Supelco®

190 L
Artikelnr. 1.02832.6190
Entzündlich
Gesundheit
Reizend
Umwelt
Anbieter:   Merck
Cyclohexan, EMPLURA®, Supelco®
Artikelnr. 1.02832.6190
Anbieter:  Merck
CAS Nummer:  110-82-7
Cyclohexan, EMPLURA®, Supelco®
Cyclohexan, EMPLURA®, Supelco®

190 L

CAS Nummer:  110-82-7
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 190 L
Product Details & Documents

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.

Synonyme:  Hexahydrobenzene, Hexamethylene
  • CAS Nummer:110-82-7
  • Formel:C₆H₁₂
  • MDL-Nummer:MFCD00003814
  • Molekulargewicht:84.16 g/mol
  • Merck-Index:13,02752
  • Siedepunkt:81 °C (1013 hPa)
  • Schmelzpunkt:7 °C
  • Dichte:0,7785 g/cm³ (20 °C)
  • Blitzpunkt:–18 °C
  • Lagertemperatur:Raumtemperatur
  • UN:1145
  • ADR:3,II
  •  :M-Clarity™ Qualitätsstufe = MQ200
  • EINECS:203-806-2

Dokumente

Zertifikate
Suche nach Chargennummer
Spezifikationen

Spezifikationen

  • VE:
    190 L

Spezifikationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.