Cyclohexan, EMPLURA®, Supelco®
Edelstahltrommel, Mehrweg, 25 L
Supplier: Merck
Jump to:
Product Details & Documents
EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.
Synonyms: Hexahydrobenzene, Hexamethylene
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Boiling Pt:81 °C (1013 hPa)
- Melting Pt:7 °C
- Density:0,7785 g/cm³ (20 °C)
- Flash Pt:–18 °C
- Storage Temperature:Raumtemperatur
- UN:1145
- ADR:3,II
- :M-Clarity™ Qualitätsstufe = MQ200
- EINECS:203-806-2
Specifications
Specifications
- Packung:Edelstahltrommel, Mehrweg
- VE:25 L



