Adipohydrazid 98%
Kst.-Flasche, 25 g
: Adipyl hydrazide
- :1071-93-8
- :C₆H₁₄N₄O₂
- :MFCD00007614
- :174.2 g/mol
- :446 °C (1013 hPa)
- :178…182 °C
- :Gefrierschrank
- :213-999-5
- :3077
- :9,III
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:178 to 182 °C
- Titration:≥97.5 %
- Packung:Kst.-Flasche
- VE:25 g



