Hydroxylammoniumchlorid 97%
Kst.-Flasche, 1 kg
Anbieter: Thermo Fisher Scientific
Hydroxylammoniumchlorid 97%
Artikelnr. ACRO170360010
Anbieter: Thermo Fisher Scientific
CAS Nummer: 5470-11-1
Springe zu:
Product Details & Documents
Synonyme: Hydroxylamine HCl, Hydroxylamine hydrochloride
- CAS Nummer:5470-11-1
- Formel:NH₂OH·HCl
- MDL-Nummer:MFCD00051089
- Molekulargewicht:69.49 g/mol
- Merck-Index:13,04853
- Siedepunkt:306 °C (1013 hPa)
- Schmelzpunkt:155…158 °C (Zers.)
- Schmelzpunkt:155…158 °C (Zersetzung)
- Dichte:1,67 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:226-798-2
- UN:2923
- ADR:8,III
Documents
Spezifikationen
Spezifikation Testergebnisse
- Appearance:White to light yellow Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:151 to 157 °C (Dec)
- Titration with NaOH:≥96.0 %
Spezifikationen
- Packung:Kst.-Flasche
- VE:1 kg
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



