Dimethyldisulfid 99%
Glasflasche für Feststoffe, 1 kg
Anbieter: Thermo Fisher Scientific
Dimethyldisulfid 99%
Artikelnr. ACRO165590010
Anbieter: Thermo Fisher Scientific
CAS Nummer: 624-92-0
Jump to:
Product Details & Documents
Synonyms: Dimethyl disulphide, Methyl disulphide
- CAS Number:624-92-0
- Formula:C₂H₆S₂
- MDL Number:MFCD00008561
- Molecular Weight:94.2 g/mol
- Boiling Pt:108…110 °C (760 mmHg)
- Melting Pt:–85 °C
- Density:1,0625 g/cm³
- Flash Pt:16 °C
- Storage Temperature:Raumtemperatur
- EINECS:210-871-0
- UN:2381
- ADR:3,II
Documents
Specifications
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5243 to 1.5263 (20°C, 589 nm)
- Water:≤0.06 %
- GC:≥98.5 %
Specifications
- Packung:Glasflasche für Feststoffe
- VE:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



