Express Eingabe
Austria
AuftragsverfolgungKontakt
Bottle pourers | Avantor
Bottle pourers
Bottle pourer, PTFE, blue
Artikelnr. COWI028.030
Anbieter:   COWIE
Bottle pourers
Artikelnr. COWI028.030
Anbieter:  COWIE
Bottle pourers
Bottle pourers
Bottle pourer, PTFE, blue
Preis
88,10 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Manufactured in pure, FDA approved, PTFE.
  • Totally chemically resistant and inert
  • To fit GL45, 38, 32 and 30 screw neck bottles
  • State cap colour required, blue, red or green
Permits direct pouring in a safe and consistent manner.
Spezifikationen
  • Bezeichnung:
    Bottle pourer, PTFE, blue
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.