Adenin 99%
Glasflasche für Feststoffe, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine
- CAS Number:73-24-5
- Formula:C₅H₅N₅
- MDL Number:MFCD00041790
- Molecular Weight:135.13 g/mol
- Merck Index:13,00152
- Melting Pt:360…365 °C (Zers.)
- Melting Pt:360…365 °C (Zersetzung)
- Density:0,99741 g/cm³ (25 °C)
- Storage Temperature:Raumtemperatur
- EINECS:200-796-1
- UN:2811
- ADR:6.1,III
Documents
ACRO14744
SDS
Specifications
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C, 5 hrs)
- Sulfated ash:≤0.1 %
- HPLC:≥98.5 %
- Titration with HClO4:≥98.5 %
Specifications
- Packung:Glasflasche für Feststoffe
- VE:100 g



