Adenin 99%
Glasflasche für Feststoffe, 25 g
: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine
- :73-24-5
- :C₅H₅N₅
- :MFCD00041790
- :135.13 g/mol
- :13,00152
- :360…365 °C (Zers.)
- :360…365 °C (Zersetzung)
- :0,99741 g/cm³ (25 °C)
- :Raumtemperatur
- :200-796-1
- :2811
- :6.1,III
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C, 5 hrs)
- Sulfated ash:≤0.1 %
- HPLC:≥98.5 %
- Titration with HClO4:≥98.5 %
- Packung:Glasflasche für Feststoffe
- VE:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



