2-Octanon 99+%
Glasflasche, 25 mL
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Synonyme: Octan-2-one
- CAS Nummer:111-13-7
- Formel:C₈H₁₆O
- MDL-Nummer:MFCD00009540
- Molekulargewicht:128.21 g/mol
- Merck-Index:13,04731
- Siedepunkt:173 °C (760 mmHg)
- Schmelzpunkt:–16 °C
- Dichte:0,819 g/cm³
- Blitzpunkt:55 °C
- Lagertemperatur:Raumtemperatur
- EINECS:203-837-1
- UN:1224
- ADR:3,III
Spezifikationen
Spezifikation Testergebnisse
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4140 to 1.4160 (20°C, 589 nm)
- GC:≥99.0 %
- Clarity of solution:Passes test
Spezifikationen
- Packung:Glasflasche
- VE:25 mL
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



