Express Eingabe
Austria
AuftragsverfolgungKontakt
1,6-Diaminohexan 99.5+%

1,6-Diaminohexan 99.5+%

Kst.-Flasche, 500 g
  ACRO120645000
 
 
1,6-Diaminohexan 99.5+%
  ACRO120645000
 :  124-09-4
1,6-Diaminohexan 99.5+%
1,6-Diaminohexan 99.5+%

Kst.-Flasche, 500 g

 :  124-09-4
 
76,40 €
   
1 * 500 g
 
 :  1,6-Hexanediamine, Hexamethylenediamine
  •  :124-09-4
  •  :C₆H₁₆N₂
  •  :MFCD00008243
  •  :116.21 g/mol
  •  :13,04713
  •  :204…205 °C (760 mmHg)
  •  :38…41 °C
  •  :0,66 g/cm³ (20 °C)
  •  :81 °C
  •  :Raumtemperatur
  •  :204-679-6
  •  :2280
  •  :8,III

Documents

 

 

  • Appearance:
    White Crystalline low melting mass (May discolor on storage)
  • Infrared spectrum:
    Conforms
  • Melting point:
    38 to 41 °C
  • Water:
    ≤0.05 % (K.F.)
  • GC:
    ≥99.50 %
  • Titration with HClO4:
    99.5 to 100.5 %

 

  • Packung:
    Kst.-Flasche
  • VE:
    500 g

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.