Docusatnatrium 96%
Kst.-Flasche, 100 g
: Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
- :577-11-7
- :C₂₀H₃₇NaO₇S
- :MFCD00012455
- :444.57 g/mol
- :12,03460
- :? 180 °C (1013 hPa)
- :153…157 °C
- :~ 1,1 g/cm³ (20 °C)
- :Raumtemperatur
- :209-406-4
- Appearance:White Waxy solid
- Infrared spectrum:Conforms
- Assay:≥95 %
- Water:≤2 %
- Color of alcohol solution:≤100 APHA (50 % solids in 1:1 alcohol/water)
- Insoluble matter:≤300 ppm (50 % in toluene)
- Iodine value:≤0.25 (on solids basis)
- Acid number:≤2.5 (on solids basis)
- Packung:Kst.-Flasche
- VE:100 g



