Docusatnatrium 96%
Kst.-Flasche, 1 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
- CAS Number:577-11-7
- Formula:C₂₀H₃₇NaO₇S
- MDL Number:MFCD00012455
- Molecular Weight:444.57 g/mol
- Merck Index:12,03460
- Boiling Pt:? 180 °C (1013 hPa)
- Melting Pt:153…157 °C
- Density:~ 1,1 g/cm³ (20 °C)
- Storage Temperature:Raumtemperatur
- EINECS:209-406-4
Specifications
Specification Test Results
- Appearance:White Waxy solid
- Infrared spectrum:Conforms
- Assay:≥95 %
- Water:≤2 %
- Color of alcohol solution:≤100 APHA (50 % solids in 1:1 alcohol/water)
- Insoluble matter:≤300 ppm (50 % in toluene)
- Iodine value:≤0.25 (on solids basis)
- Acid number:≤2.5 (on solids basis)
Specifications
- Packung:Kst.-Flasche
- VE:1 kg



