Methyloleat, technical
Glasflasche für Feststoffe, 500 g
Anbieter: Thermo Fisher Scientific
Methyloleat, technical
Artikelnr. ACRO414645000
Anbieter: Thermo Fisher Scientific
CAS Nummer: 67762-38-3
Jump to:
Product Details & Documents
Synonyms: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester
- CAS Number:67762-38-3
- Formula:C₁₉H₃₆O₂
- MDL Number:MFCD00009578
- Molecular Weight:296.49 g/mol
- Merck Index:12,06965
- Boiling Pt:186 °C (9 mmHg)
- Melting Pt:–20 °C
- Density:0,87 g/cm³
- Flash Pt:170 °C
- Storage Temperature:Kühlschrank
- EINECS:203-992-5
Documents
ACRO41464
SDS
Specifications
Specification Test Results
- Appearance:Clear yellow to amber Viscous liquid
- Infrared spectrum:Conforms
- GC:≥55 %
- HPLC:≥98.0 %
Specifications
- Packung:Glasflasche für Feststoffe
- VE:500 g



