Hydroxylammoniumchlorid ACS
Kst.-Flasche, 100 g
Anbieter: Thermo Fisher Scientific
Hydroxylammoniumchlorid ACS
Artikelnr. ACRO412051000
Anbieter: Thermo Fisher Scientific
CAS Nummer: 5470-11-1
Springe zu:
Product Details & Documents
Synonyme: Hydroxylamine HCl, Hydroxylamine hydrochloride
- CAS Nummer:5470-11-1
- Formel:NH₂OH·HCl
- MDL-Nummer:MFCD00051089
- Molekulargewicht:69.49 g/mol
- Merck-Index:13,04853
- Siedepunkt:306 °C (1013 hPa)
- Schmelzpunkt:155…158 °C
- Dichte:1,67 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:226-798-2
- UN:2923
- ADR:8,III
Dokumente
Spezifikationen
Spezifikation Testergebnisse
- Appearance:White Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Residue after ignition:≤0.05 %
- Titration with KMnO4:≥96.0 %
- Clarity of alcohol solution:Passes test
- Ammonium (NH4):Passes test
- Iron (Fe):≤5 ppm
- Heavy metals (ICP-OES):≤5 ppm
- Sulfur compounds (as SO4):≤0.005 %
- Titrable free acid:≤0.25 meq/g
Spezifikationen
- Packung:Kst.-Flasche
- VE:100 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



