AuftragsverfolgungKontakt
Torso | Avantor
Torso
Torso (Electronic Light Contr.)
Artikelnr. BINH1005618
Marketsource
Anbieter:   3B Scientific
Torso
Artikelnr. BINH1005618
Anbieter:  3B Scientific
Marketsource
Torso
Torso
Torso (Electronic Light Contr.)
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
This easy to transport torso allows the practice of adult CPR techniques. Optionally child CPR can also be practiced by changing the supplied springs inside the torso. Individual student face masks with a one-way non-rebreathing valve and disposable airways provide maximum protection against cross-contamination.
  • With light controller
Spezifikationen
  • Bezeichnung:
    Torso (Electronic Light Contr.)