Büchner-Trichter
175 mm, 4000 ml, 4
Artikelnr. 511-0784
Anbieter: SciLabware
Springe zu:
Product Details & Documents
PYREX®, borosilicate glass, with sintered glass disc.
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Very good temperature resistance
For convenient filtration of solutions.
Spezifikationen
- Platte Ø:175 mm
- Porosität:4
- Kapazität:4000 ml
- Seq:56



