Xanthydrol 97+%
Kst.-Flasche, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 9-Hydroxyxanthene, 9-Xanthenol, 9H-Xanthen-9-ol
- CAS Number:90-46-0
- Formula:C₁₃H₁₀O₂
- MDL Number:MFCD00005057
- Molecular Weight:198.22 g/mol
- Boiling Pt:338 °C (1013 hPa)
- Melting Pt:124…127 °C
- Storage Temperature:Kühlschrank
- EINECS:201-996-1
Specifications
Specification Test Results
- Appearance:White to almost white Powder
- Infrared spectrum:Conforms
- Melting point:121 to 126 °C
- HPLC:≥97.0 %
Specifications
- Packung:Kst.-Flasche
- VE:25 g



