Xanthydrol 97+%
Glasflasche für Feststoffe, 5 g
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Synonyme: 9-Hydroxyxanthene, 9-Xanthenol, 9H-Xanthen-9-ol
- CAS Nummer:90-46-0
- Formel:C₁₃H₁₀O₂
- MDL-Nummer:MFCD00005057
- Molekulargewicht:198.22 g/mol
- Siedepunkt:338 °C (1013 hPa)
- Schmelzpunkt:124…127 °C
- Lagertemperatur:Kühlschrank
- EINECS:201-996-1
Documents
ACRO27152
SDS
Spezifikationen
Spezifikation Testergebnisse
- Appearance:White to almost white Powder
- Infrared spectrum:Conforms
- Melting point:121 to 126 °C
- HPLC:≥97.0 %
Spezifikationen
- Packung:Glasflasche für Feststoffe
- VE:5 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



