AuftragsverfolgungKontakt
Rückschlagventile
6,4 - 9,5 mm ID
Catalog # BELAH197160000
Rückschlagventile
Catalog # BELAH197160000
Rückschlagventile
Rückschlagventile
6,4 - 9,5 mm ID
Price
145,00 €
Log in for availability
1 * 6 ST
Product Details & Documents
PP valve, translucent with neoprene valve disk. Stops reverse flow of liquid when pressure is interrupted.
  • Useful when connecting a vacuum line to a filter flask
  • Recommended maximum pressure 20 psi. (1034 Torr)
  • Autoclavable
Specifications
  • Schlauchgröße:
    6,4 - 9,5 mm ID