Heating tapes, TeMS2
Heating tape TeMS2, 22,2 m length, 100 W
Artikelnr. ISOP411376-000
Anbieter: ISOPAD
Springe zu:
Product Details & Documents
Isopad IT-TeMS is a preterminated and PTFE insulated flexible heating tape.
- Temperature resistant up to 260 °C
- Non hazardous and used in ordinary areas
- Parallel arrangement ensures a wide heat transfer surface
IT-TeMS2 is used in applications ranging from simple frost protection to temperature maintenance on short pipework and apparatus as well as for general container heating applications. TeMS4 or TeMS6 tapes can be used when more power is required.
Spezifikationen
- Bezeichnung:Heating tape TeMS2, 22,2 m length, 100 W
- Seq:02



