Express Eingabe
Austria
AuftragsverfolgungKontakt
Organic acids solution kit

Organic acids solution kit

Organic acids test sample, 20 ml, 20 mL
Artikelnr. HEWL8500-6900
Anbieter:   Agilent
Organic acids solution kit
Artikelnr. HEWL8500-6900
Anbieter:  Agilent
CAS Nummer:  
Organic acids solution kit

Organic acids test sample, 20 ml, 20 mL

CAS Nummer:  
Preis
336,00 €
Log in for availability
1 * 20 mL
Product Details & Documents

The Organic Acids Solutions Kit contains capillaries, buffers, an organic acids standard and instructions complete with a sample electropherogram to help you begin organic acids analyses immediately.

  • Ideal for the analysis of short alkyl-chain carboxylic acids
  • Analyze food applications, including beer, wine and juice
  • Use for chemical applications, including raw materials and drug formulations

Employing an indirect UV detection agent optimized for organic acids, the methodology is simple, sensitive, and provides accurate quantitative analysis. Suited for the analysis of organic acids in a wide range of matrices, it is especially useful for determination of organic acids in beverages, and food.

Specifications

Specifications

  • VE:
    20 mL
  • Bezeichnung:
    Organic acids test sample, 20 ml

Specifications