Solvent filter/degasser
HPLC solvent filter/degasser assembly, PTFE
Artikelnr. HEWL3150-0577
Anbieter: Agilent
Springe zu:
Product Details & Documents
It prevents the formation of spurious peaks within the detector due to solvent outgassing at the low-pressure end of the chromatograph.
- Degasses eluents as particulates are removed
- Increases solvent inlet lifetime
- Eliminates pump downtime caused by air locks and particulates in check valves
- Decreases piston wear, while increasing column life
Spezifikationen
- Bezeichnung:HPLC solvent filter/degasser assembly
- Material:PTFE
- Seq:03