Express Eingabe
Austria
AuftragsverfolgungKontakt
Split/splitless inlet seals | Avantor
Split/splitless inlet seals
Septum retainer nut for headspace
Artikelnr. HEWL18740-60830
Anbieter:   Agilent
Split/splitless inlet seals
Artikelnr. HEWL18740-60830
Anbieter:  Agilent
Split/splitless inlet seals
Split/splitless inlet seals
Septum retainer nut for headspace
Preis
253,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
These inlet seals have been developed to ensure that you have a consistent and inert surface to properly seal the inlet and prevent sample degradation.
  • Very reproducible smooth surface, eliminating traces of machining grooves that can be the source of minute leaks
  • High quality, leak-free seal
Lieferumfang: An inlet washer is provided with each inlet seal.
Spezifikationen
  • Bezeichnung:
    Septum retainer nut for headspace
  • Seq:
    08
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.