AuftragsverfolgungKontakt
Analogue stopwatches, split-second addition timer | Avantor
Analogue stopwatches, split-second addition timer
Analoge Stoppuhr, Hebel, 7 Steine, 1/100 Minuten 30 Minuten, 130 g, 55 mm
Artikelnr. HANH135.0201-0S
Anbieter:   HANHART
Analogue stopwatches, split-second addition timer
Artikelnr. HANH135.0201-0S
Anbieter:  HANHART
Analogue stopwatches, split-second addition timer
Analogue stopwatches, split-second addition timer
Analoge Stoppuhr, Hebel, 7 Steine, 1/100 Minuten 30 Minuten, 130 g, 55 mm
Preis
714,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Addition timer for time measurement with interrupt option, also drag indicator for split-time measurement.
  • Chrome plated brass casing, diamond cut
  • Lever, 7 jewels
  • Shock resistant, additional dust and water protection
Spezifikationen
  • Ø:
    55 mm
  • Bezeichnung:
    Analoge Stoppuhr, Hebel, 7 Steine, 1/100 Minuten 30 Minuten, 130 g