Express Eingabe
Austria
AuftragsverfolgungKontakt
Crown stopwatches

Crown stopwatches

Analoge Stoppuhr, 1/5 Sekunden, 30 Minuten, 120 g, Verchromt, 55 mm
Catalog # 609-1119
Supplier:   HANHART
Crown stopwatches
Catalog # 609-1119
Supplier:  HANHART
Crown stopwatches
Crown stopwatches

Analoge Stoppuhr, 1/5 Sekunden, 30 Minuten, 120 g, Verchromt, 55 mm

Price
384,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Chrome plated and polished brass casing, diamond cut.

  • Pin lever, seven jewels
  • Protected against shock, dust and water
  • Industry model: Additional case seals to give extra protection against dust and water
Specifications
  • Ø:
    55 mm
  • Farbe:
    Verchromt
  • Bezeichnung:
    Analoge Stoppuhr, 1/5 Sekunden, 30 Minuten, 120 g