Laboratory forceps, blunt
Straight, blunt serrated tips, 140 mm
232-1920
Wironit steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).
- Resistant to corrosion
- Anti-magnetic
- Typ:Straight, blunt serrated tips
- Länge:140 mm
Wironit steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).