AuftragsverfolgungKontakt
Electrolyte Solutions
Electrolyte Solutions
Elektrolyt 1M KNO3, Silber-frei, 100 ml, 100 ml
Catalog # 620-4934
Supplier:   HAMILTON BONADUZ
Electrolyte Solutions
Catalog # 620-4934
Supplier:  HAMILTON BONADUZ
CAS Number:  
Electrolyte Solutions
Electrolyte Solutions
Elektrolyt 1M KNO3, Silber-frei, 100 ml, 100 ml
CAS Number:  
Price
32,20 €
Log in for availability
1 * 100 mL
Product Details & Documents
A variety of electrolyte solutions for pH and ORP electrodes.
Documents
Specifications
Specifications
  • VE:
    100 ml
  • Bezeichnung:
    Elektrolyt 1M KNO3, Silber-frei, 100 ml