Microplates, 96-Well
U-Boden, transparent, 40 - 280 µl, 96, -, 323 µl
Anbieter: Greiner Bio-One
Springe zu:
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Dokumente
Spezifikationen
- Anz. Wells:96
- Empf. Arbeitsvolumen:40 - 280 µl
- Steril:-
- Bezeichnung:U-Boden, transparent
- Seq:03
- Gesamtvolumen:323 µl