Capcell Pak HPLC Columns
Analytisch, SCX UG 80, Edelstahl, 80 Å, 5 µm, 4,60 mm, 100 mm
PHECCH0-9312
New Product
- Säulenform:Analytisch
- Ø int.:4,60 mm
- Partikelgröße:5 µm
- Porengröße:80 Å
- Länge:100 mm
- Seq:7
- Säulenmaterial:Edelstahl
- Phase:SCX UG 80
- Cat. No.:PHECCH0-9312
- pH-Bereich:2,0 - 7,5
- Endcapped:Ja
- Kohlenstoffbelastung:0%