4-Hydroxyzimtsäure 98%
Bottle
APOSOR21362-25G
New Product
: p-Coumaric acid, p-Cumaric acid, 3-(4-Hydroxyphenyl)acrylic acid, p-Hydroxy-Cinnamic Acid
- :7400-08-0
- :C₉H₈O₃
- :MFCD00004399
- :164.16 g/mol
- :346 °C
- :212…214 °C
- :Raumtemperatur
- Packung:Bottle